Skip to Content
Merck

Skip To

SRP5085

SIK (1-303), active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MSK, SIK1, SNF1LK

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Specific activity:
296-401 nmol/min·mg
Assay:
≥70% (SDS-PAGE)
Recombinant:
expressed in baculovirus infected Sf9 cells


recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

296-401 nmol/min·mg

mol wt

~36 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SIK1(150094)

General description

SIK is a protein kinase that is involved in regulating AMPK-related kinases. SIK may mediate the physiological effects of LKB1, including its tumour suppressor function. SIK is also involved in signaling by various proteins like STRAD, NUAK1, NUAK2, BRSK1, BRSK2, QIK, QSK, SIK, MARK1, MARK2, MARK3, MARK4 and MELK that are related to AMPK. Activation of SIK1 by phosphorylation on thr322 can lead to an increase in the catalytic activity of sodium/potassium ATPase alpha subunit at the plasma membrane. This results in an increase in intracellular sodium in intact mammalian cells.

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SRP5086SRP5040SRP5047
Gene Information

human ... SIK1(150094)

Gene Information

human ... SLK(9748)

Gene Information

human ... CHUK(1147)

Gene Information

human ... Map3K1(4214)

assay

≥70% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

specific activity

296-401 nmol/min·mg

specific activity

3.9-5.3 nmol/min·mg

specific activity

2.1-2.9 nmol/min·mg

specific activity

697-943 nmol/min·mg

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in baculovirus infected Sf9 cells

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

288-32-4

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Hiroshi Takemori et al.
The FEBS journal, 274(13), 3202-3209 (2007-06-15)
The transcription factor cAMP response element-binding protein (CREB) plays important roles in gene expression induced by cAMP signaling and is believed to be activated when its Ser133 is phosphorylated. However, the discovery of Ser133-independent activation by the activation of transducer
Mattias Sjöström et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(43), 16922-16927 (2007-10-18)
In mammalian cells, active sodium transport and its derived functions (e.g., plasma membrane potential) are dictated by the activity of the Na(+),K(+)-ATPase (NK), whose regulation is essential for maintaining cell volume and composition, as well as other vital cell functions.



Global Trade Item Number

SKUGTIN
SRP5085-10UG04061832942131

Questions

Reviews

No rating value

Active Filters