SRP0463
BRD4, BD1, BD2 (49-460) GST tag human
recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Synonym(s):
CAP, HUNK1, HUNKI, MCAP, bromodomain containing 4
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥80% (SDS-PAGE)
form
aqueous solution
mol wt
73.3 kDa
packaging
pkg of 100 μg
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... BRD4(23476)
General description
BRD4 (bromodomain-containing protein 4) belongs to the BET (bromodomain and extra-terminal) family. The protein has two bromodomains (BD1 and BD2) connected via a long interdomain linker.
Human Bromodomain containing 4, also known as BRD4, HUNK1, or MCAP (GenBank Accession No. NM_ NM_058243) amino-acids 49-460 with N-terminal GST-tag, MW = 73.3kDa, expressed in an Escherichia coli expression system.
Human Bromodomain containing 4, also known as BRD4, HUNK1, or MCAP (GenBank Accession No. NM_ NM_058243) amino-acids 49-460 with N-terminal GST-tag, MW = 73.3kDa, expressed in an Escherichia coli expression system.
Biochem/physiol Actions
BRD4 (bromodomain-containing protein 4) works as a reader of epigenetic marks and is involved in chromatin remodeling and transcriptional regulation. Therefore, it is a target for oncology, inflammation and cardiovascular disease. This gene is upregulated in hepatocellular carcinoma and is associated with poor prognosis. BRD4 is also associated with melanoma, hepatocellular carcinoma, multiple myeloma, Burkitt′s lymphoma, acute myeloid leukemia and breast cancer. BRD4 connects cell cycle and transcription, works as a scaffold for transcription factors and bookmarks active genes during mitosis. It interacts with acetylated histone-3 and histone-4 tails and helps in maintaining chromatin architecture. Also, it regulates transcription elongation via phosphorylation of RNA polymerase II.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Segmental Isotope Labelling of an Individual Bromodomain of a Tandem Domain BRD4 Using Sortase A.
PLoS ONE, 11, e0154607-e0154607 (2016)
BRD4 promotes tumor growth and epithelial-mesenchymal transition in hepatocellular carcinoma.
International Journal of Immunopathology and Pharmacology, 28, 36-36 (2015)
High level of BRD4 promotes non-small cell lung cancer progression.
Oncotarget, 7, 9491-9491 (2016)
Journal of molecular medicine (Berlin, Germany), 90(5), 587-595 (2011-11-29)
The bromodomain protein BRD4 is involved in cell proliferation and cell cycle progression, primarily through its role in acetylated chromatin-dependent regulation of transcription at targeted loci. Here, we show that BRD4 is frequently downregulated by aberrant promoter hypermethylation in human
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service