Skip to Content
Merck
All Photos(2)

Documents

SML3735

Sigma-Aldrich

DRAQ5

≥98% (HPLC)

Synonym(s):

1,5-Bis[[2-(dimethylamino)ethyl]amino]-4,8-dihydroxy-9,10-anthracenedione, 1,5-Bis[[2-(dimethylamino)ethyl]amino]-4,8-dihydroxyanthracene-9,10-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28N4O4
CAS Number:
Molecular Weight:
412.48
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, White to very dark brown

solubility

soluble (DMSO: 1M HCl 2 mg/mL clear)

storage temp.

2-8°C

Biochem/physiol Actions

Nucleated cells detecting fluorochrome; DNA probe.


DRAQ5 is a deep red fluorescing anthraquinone dye that displays high affinity for DNA and a high capacity to rapidly enter living cells or stain fixed cells (λex = 647 nm, λem = 665–780 nm).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear cytometry and chromatin organization
Cytometry. Part A : the Journal of the International Society For Analytical Cytology, 93(8), 771-784 (2018)
Assessment of photoacoustic tomography contrast for breast tissue imaging using 3D correlative virtual histology
Scientific Reports, 12(1), 2532-2532 (2022)
Alena Gschwind et al.
Cellular & molecular biology letters, 27(1), 32-32 (2022-04-07)
Autophagy plays an essential role in maintaining cellular homeostasis and in the response to cellular stress. Autophagy is also involved in cell cycle progression, yet the relationship between these processes is not clearly defined. In exploring this relationship, we observed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service