Skip to Content
Merck

Skip To

SML2745

GSK′481

≥98% (HPLC)

Synonym(s):

(S)-5-Benzyl-N-(5-methyl-4-oxo-2,3,4,5-tetrahydrobenzo[b]-[1,4]oxazepin-3-yl)isoxazole-3-carboxamide, GSK 2882481, GSK 481, GSK-2882481, GSK-481, GSK2882481, GSK481

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
CZK 3,380.00
CZK 2,873.00
25 mg
Please contact Customer Service for Availability
CZK 12,700.00
CZK 10,795.00

About This Item

Empirical Formula (Hill Notation):
C21H19N3O4
CAS Number:
Molecular Weight:
377.39
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder

CZK 2,873.00

List PriceCZK 3,380.00Save 15%

Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

N1(c2c(cccc2)OC[C@@H](C1=O)NC(=O)c3n[o]c(c3)Cc4ccccc4)C

InChI

1S/C21H19N3O4/c1-24-18-9-5-6-10-19(18)27-13-17(21(24)26)22-20(25)16-12-15(28-23-16)11-14-7-3-2-4-8-14/h2-10,12,17H,11,13H2,1H3,(H,22,25)/t17-/m0/s1

InChI key

KNOUWGGQMADIBV-KRWDZBQOSA-N

Biochem/physiol Actions

A highly selective ATP site-targeting inhibitor against human/monkey, but not non-primate, RIP1.
GSK′481 is a highly potent receptor interacting protein 1 kinase (RIP1, RIPK1) inhibitor (human/monkey RIP1 IC50 = 1.6/2.5 nM; [ATP] = 50 μM) that targets RIP1 ATP-binding pocket with high affinity (kon = 0.66/μM/sec, t1/2 = 11 min) with little potency toward non-primate RIP1 (IC50 = 2.0/3.2/6.3/7.9/>10 μM against rabbit/mouse/rat/dog/minipig RIP1) and no off-target affinity when profiled at 10 μM by a 456-kinase panel. GSK′481 blocks caspase inhibitor/TNF-induced necroptosis in human monocytic U937 cultures (IC50 = 10 nM) and inhibits S166 autophosphorylation of exogenously expressed human, but not mouse, RIP1 in HEK293T transfectants (IC50 = 2.8 nM and >10 μM, respectively).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2376SML2377SML2770
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

white to beige


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
SML2745-5MG04061841380856
SML2745-25MG04061841380849

Questions

Reviews

No rating value

Active Filters