Skip to Content
Merck

Skip To

SML0638

S0859

≥98% (HPLC)

Synonym(s):

2-Chloro-N-[[2′-[(cyanoamino)sulfonyl][1,1′-biphenyl]-4-yl]methyl]-N-[(4-methylphenyl)methyl]-benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
3 070,00 CZK
25 mg
Please contact Customer Service for Availability
12 400,00 CZK

About This Item

Empirical Formula (Hill Notation):
C29H24ClN3O3S
CAS Number:
Molecular Weight:
530.04
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

3 070,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(NC#N)c1c(cccc1)c2ccc(cc2)CN(Cc4ccc(cc4)C)C(=O)c3c(cccc3)Cl

InChI

1S/C29H24ClN3O3S/c1-21-10-12-22(13-11-21)18-33(29(34)26-7-2-4-8-27(26)30)19-23-14-16-24(17-15-23)25-6-3-5-9-28(25)37(35,36)32-20-31/h2-17,32H,18-19H2,1H3

InChI key

ITDBPOSLOROLMT-UHFFFAOYSA-N

Biochem/physiol Actions

S0859 is a selective high-affinity generic inhibitor of the Na+/HCO3- sodium bicarbonate co-transporter (NBC).
S0859 is a selective high-affinity generic inhibitor of the Na+/HCO3- sodium bicarbonate co-transporter (NBC). S0859 does not inhibit Na+-H+ exchange (NHE).
S0859 helps to prevent the depolarization-induced alkalinization (DIA) in astrocytes.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0853SML2657SML1510
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 10 mg/mL, clear

solubility

H2O: 1 mg/mL, clear (warmed)

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 5 mg/mL, clear (warmed)

color

white to light brown

color

white to beige

color

white to beige

color

white to beige


Hazard Codes

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters