Skip to Content
Merck
All Photos(1)

Key Documents

SML0222

Sigma-Aldrich

TVP1022

≥98% (HPLC)

Synonym(s):

(1S)-2,3-Dihydro-N-2-propyn-1-yl-1H-inden-1-amine, (S)-Rasagiline, S-(-)-N-Propargyl-1-aminoindan

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CZK 2,680.00
25 MG
CZK 10,300.00

CZK 2,680.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CZK 2,680.00
25 MG
CZK 10,300.00

About This Item

Empirical Formula (Hill Notation):
C12H13N
CAS Number:
Molecular Weight:
171.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

CZK 2,680.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

(Powder, Film or Oil)

solubility

DMSO: ≥15 mg/mL

storage temp.

2-8°C

SMILES string

C#CCN[C@H]1CCc2ccccc12

InChI

1S/C12H13N/c1-2-9-13-12-8-7-10-5-3-4-6-11(10)12/h1,3-6,12-13H,7-9H2/t12-/m0/s1

InChI key

RUOKEQAAGRXIBM-LBPRGKRZSA-N

Biochem/physiol Actions

TVP1022 is an S-isomer of rasagiline, a PKC Pathway activator and neuroprotective and cytoprotective
TVP1022, the S-isomer of rasagiline, an anti-Parkinson drug, appears to have the same neuroprotective activity as the R-isomer, but is 1000-fold less active as an MAO-B inhibitor. TVP1022, a neuroprotective and cytoprotective molecule, is also cardioprotective in rat models. Its activity is believed to involve stabilization of mitochondrial membrane potential, induction of Bcl-2 and activation of the PKC signalling pathway, enhancing the phosphorylation of protein kinase C and glycogen synthase kinase-3β.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service