Skip to Content
Merck

Skip To

SML0104

Ascochlorin

≥98% (HPLC), from Verticillium hemipterigenum

Synonym(s):

5-Chloro-2,4-dihydroxy-6-methyl-3-[(2E,4Z)-3-methyl-5-(1,2,6-trimethyl-3-oxocyclohexyl)penta-2,4-dienyl]benzaldehyde, Antibiotics LL-Z 1272g, Ilicicolin D, NSC 287492

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C23H29ClO4
CAS Number:
Molecular Weight:
404.93
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)


biological source

Verticillium hemipterigenum

assay

≥98% (HPLC)

solubility

DMSO: soluble, H2O: insoluble, chloroform: soluble, ethyl acetate: soluble, methanol: soluble

antibiotic activity spectrum

viruses

mode of action

enzyme | inhibits

storage temp.

−20°C

InChI

1S/C23H29ClO4/c1-13(10-11-23(5)14(2)7-9-19(26)16(23)4)6-8-17-21(27)18(12-25)15(3)20(24)22(17)28/h6,10-12,14,16,27-28H,7-9H2,1-5H3/b11-10+,13-6+

InChI key

SETVRSKZJJWOPA-OGCXVWDESA-N

Application

The effect of Ascochlorin on MMP-9 was compared with that of Silibinin in MCF-7 human breast carcinoma cells.[1]

Biochem/physiol Actions

Ascochlorin is a prenyl-phenol compound and a dihydroorotate dehydrogenase (DHODH) inhibitor. It has hypolipidemic functionality and suppresses hypertension as well as immune responses.[2] It is a glycoside, which also elicits inhibition of the respiratory chain cytochrome bc1.[3]
Ascochlorin is an isoprenoid antibiotic initially identified as an antiviral and antitumor agent.
Ascochlorin is an isoprenoid antibiotic produced by Verticillium hemipterigenum, initially identified as an antiviral and antitumor agent. Research also indicates that ascochlorin inhibits the Qi and Qo quinone binding sites of the mitochondria cytochrom bc1 complex. Moreover, ascochlorin activates p53, probably as a result of its inhibitory effect on mitochondrial respiration. In addition, ascochlorin has the ability to suppress the nuclear subscription enzyme protein-1, a nuclear transcription factor activator, which leads to suppression of the extra-cellular enzyme, matrix metalloproteinase-9 (MMP9). The regulation of MMP is implicated in renal development, macrophage differentiation, atherosclerosis, inflammation, rheumatoid arthritis, and tumor invasion. Ascochlorin was also found to be effective against Mx-1, an estrogen lacking breast cancer cell line.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters