Skip to Content
Merck

Skip To

SMB01072

Turmeronol A

≥90% (LC/MS-ELSD)

Synonym(s):

(6S)-2-Methyl-6-(3-hydroxy-4-methylphenyl)-2-hepten-4-one, (S)-6-(3-Hydroxy-4-methylphenyl)-2-methylhept-2-en-4-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Please contact Customer Service for Availability
8 470,00 CZK
6 945,40 CZK

About This Item

Empirical Formula (Hill Notation):
C15H20O2
CAS Number:
Molecular Weight:
232.32
UNSPSC Code:
12352205
NACRES:
NA.25

6 945,40 CZK

List Price8 470,00 CZKSave 18%

Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

plant

assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

232.32

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

Oc1c(ccc(c1)[C@H](CC(=O)C=C(C)C)C)C

InChI

1S/C15H20O2/c1-10(2)7-14(16)8-12(4)13-6-5-11(3)15(17)9-13/h5-7,9,12,17H,8H2,1-4H3/t12-/m0/s1

InChI key

OSIFVLKZUWRNBN-LBPRGKRZSA-N

General description

Turmeronol, a phenolic sesquiterpenoid, is a natural product commonly available from the rhizomes of Curucuma longa L. (Turmeric). This bioactive metabolite derived from plant source exhibits various biological activities such as anti-inflammatory, antioxidant, anticancer, and antimicrobial properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Turmeronol A is inhibitor of soybean lipoxygenase at the IC50 value of 16 μM and prevents the autoxidation of linoleic acid at a concentration of ca. 200 ppm.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SMB01039SMB00232SMB00138
description

≥90% (LC/MS-ELSD)

description

≥90% (LC/MS-ELSD)

description

≥95% (LC/MS-ELSD)

description

≥95% (LC/MS-ELSD)

assay

≥90% (LC/MS-ELSD)

assay

≥90% (LC/MS-ELSD)

assay

≥95% (LC/MS-ELSD)

assay

≥95% (LC/MS-ELSD)

biological source

plant

biological source

plant

biological source

-

biological source

-

form

solid

form

solid

form

solid

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

mol wt

232.32

mol wt

211.26

mol wt

-

mol wt

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters