Skip to Content
Merck
All Photos(1)

Key Documents

SMB00232

Sigma-Aldrich

Clusiacitran A

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CZK 10,800.00

CZK 10,800.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
CZK 10,800.00

About This Item

Empirical Formula (Hill Notation):
C23H24O4
CAS Number:
Molecular Weight:
364.43
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

CZK 10,800.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC1(C)Oc2c3C4CC(C)(CCC14)Oc3cc(O)c2C(=O)c5ccccc5

InChI

1S/C23H24O4/c1-22(2)15-9-10-23(3)12-14(15)18-17(26-23)11-16(24)19(21(18)27-22)20(25)13-7-5-4-6-8-13/h4-8,11,14-15,24H,9-10,12H2,1-3H3

InChI key

XUPOZHDUFRMVTF-UHFFFAOYSA-N

General description

Natural product derived from plant source.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service