Skip to Content
Merck

Skip To

S9995

Recombinant SARS-CoV-2 nucleocapsid protein

Synonym(s):

2019-nCoV, SARS CoV2 NP, Severe acute respiratory syndrome coronavirus 2, coronavirus Nucleoprotein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Please contact Customer Service for Availability
CZK 10,600.00

About This Item

NACRES:
NA.26
UNSPSC Code:
12352202
Technique(s):
ELISA: suitable, lateral flow assay: suitable
Species reactivity:
-
Application:
ELISA, lateral flow assay
Citations:
-

CZK 10,600.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Escherichia coli

Quality Segment

assay

95% (SDS-PAGE)

form

frozen liquid

mol wt

~48 kDa

contains

0.025% sodium azide

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.5 mg/mL protein (Lowry method)

technique(s)

ELISA: suitable, lateral flow assay: suitable

storage temp.

−70°C

General description

This full-length SARS-CoV-2 virus nucleoprotein protein gene was cloned, expressed and purified from E.coli. The nucleocapsid protein is quite conserved. Using nucleocapsid protein for acute / early antibodies such as IgA/ IgM should make it possible to detect antibodies directed against the variants of concern in a satisfactory way at an early stage of illness.

Physical form

Frozen liquid in PBS and 25mM K2CO3

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABF2787MABF2788SAB3501124
biological source

Escherichia coli

biological source

mouse

biological source

mouse

biological source

rabbit

mol wt

~48 kDa

mol wt

calculated mol wt 45.63 kDa, observed mol wt ~53 kDa

mol wt

calculated mol wt 45.63 kDa, observed mol wt ~50 kDa

mol wt

-

form

frozen liquid

form

-

form

-

form

liquid

storage temp.

−70°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

technique(s)

ELISA: suitable, lateral flow assay: suitable

technique(s)

ELISA: suitable, immunofluorescence: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

ELISA: suitable, immunofluorescence: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

ELISA: suitable, western blot: suitable

concentration

>0.5 mg/mL protein (Lowry method)

concentration

-

concentration

-

concentration

1 mg/mL


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

nwg



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
S9995-1MG04065266408072
S9995-100UG04065266408065

Questions

Reviews

No rating value

Active Filters