Skip to Content
Merck
All Photos(1)

Key Documents

S7323

Sigma-Aldrich

14-3-3 Sigma histidine-tagged human

recombinant, expressed in E. coli, C-terminal, ≥90% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

14-3-3σ, Epitheliial cell marker protein 1, HME1, Makorin, ring finger protein 3, Mkrn3, Mmel, SFN, Stratifin, Tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, sigma polypeptide, YWHAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~30 kDa (14-3-3 proteins)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SFN(2810)

Biochem/physiol Actions

14-3-3 proteins are involved in a multitude of biological processes and play a regulatory role in processes such as apoptotic cell death, mitogenic signal transduction, and cell cycle control.[1] 14-3-3σ proteins play a critical role in signal transduction pathways and cell cycle regulation. 14-3-3σ is the only 14-3-3 isoform induced by the tumor suppressor protein p53, in response to γ-irradiation and other DNA-damaging agents.[2] 14-3-3σ is a p53-regulated inhibitor of G2/M progression[3] and acts as a tumor suppressor gene that is inactivated by methylation of its 5′ CpG islands in epithelial tumor cells.

Physical form

Supplied as a 0.2 μm filtered solution in 50 mM Tris-HCl, pH 7.6, 1 mM EDTA, 1 mM DTT, and 20% glycerol (w/v).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huiling Yang et al.
Cancer research, 66(6), 3096-3105 (2006-03-17)
14-3-3 sigma is induced by tumor suppressor protein p53 in response to DNA damage. p53 can directly transactivate the expression of 14-3-3 sigma to cause a G(2) cell cycle arrest when cell DNA is damaged. The expression of 14-3-3 sigma
José M A Moreira et al.
Molecular & cellular proteomics : MCP, 7(7), 1225-1240 (2008-04-02)
The 14-3-3 proteins constitute a family of highly conserved and broadly expressed multifunctional polypeptides that are involved in a variety of important cellular processes that include cell cycle progression, growth, differentiation, and apoptosis. Although the exact cellular function(s) of 14-3-3
H Hermeking et al.
Molecular cell, 1(1), 3-11 (1998-07-11)
Exposure of colorectal cancer (CRC) cells to ionizing radiation results in a cell-cycle arrest in G1 and G2. The G1 arrest is due to p53-mediated induction of the cyclin-dependent kinase inhibitor p21WAF1/CIP1/SDI1, but the basis for the G2 arrest is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service