Skip to Content
Merck

Skip To

S2382

Sera from sheep

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 mL
Please contact Customer Service for Availability
CZK 784.00

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207
Biological source:
sheep
Origin:
USA origin
Form:
lyophilized powder

CZK 784.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

sheep

form

lyophilized powder

origin

USA origin

storage temp.

2-8°C

Application

Sheep serum is useful for the propagation of long-term cultures of erythrocytic stages of the Theileria uilenbergi intracellular protozoan parasite.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I5131A3264SAB3701358
biological source

sheep

biological source

sheep

biological source

sheep

biological source

sheep

origin

USA origin

origin

-

origin

-

origin

-

form

lyophilized powder

form

essentially salt-free, lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Joana P G Miranda et al.
American journal of veterinary research, 67(11), 1908-1913 (2006-11-03)
To establish optimal conditions for long-term culture of the erythrocytic stage of Theileria uilenbergi. Red blood cells from 3 splenectomized sheep experimentally infected with a blood stabilate of T uilenbergi. Cultures of T uilenbergi were initiated by use of blood



Global Trade Item Number

SKUGTIN
S2382-1ML04061836918798
S2382-5ML04061836918811

Questions

Reviews

No rating value

Active Filters