Skip to Content
Merck
All Photos(2)

Documents

R140

Sigma-Aldrich

Ro 04-6790 dihydrochloride

solid

Synonym(s):

4-Amino-N-[2,6-bis(methylamino)-4-pyrimidinyl]benzenesulfonamide dihydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N6O2S · 2HCl
CAS Number:
Molecular Weight:
381.28
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

DMSO: >10 mg/mL
H2O: >10 mg/mL

originator

Roche

SMILES string

Cl.Cl.CNc1cc(NS(=O)(=O)c2ccc(N)cc2)nc(NC)n1

InChI

1S/C12H16N6O2S.2ClH/c1-14-10-7-11(17-12(15-2)16-10)18-21(19,20)9-5-3-8(13)4-6-9;;/h3-7H,13H2,1-2H3,(H3,14,15,16,17,18);2*1H

InChI key

QOAXXMSJHQHSFV-UHFFFAOYSA-N

Gene Information

human ... HTR6(3362)

Biochem/physiol Actions

Selective 5-HT6 serotonin receptor antagonist.

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Caution

Photosensitive

Legal Information

Manufactured and sold under license from Hoffman LaRoche.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Sleight et al.
British journal of pharmacology, 124(3), 556-562 (1998-07-01)
1. This study describes the in vitro characterization of two potent and selective 5-HT6 receptor antagonists at the rat and human recombinant 5-HT6 receptor. 2. In binding assays with [3H]-LSD, 4-amino-N-(2,6 bis-methylamino-pyrimidin-4-yl)-benzene sulphonamide (Ro 04-6790) and 4-amino-N-(2,6 bis-methylamino-pyridin-4-yl)-benzene sulphonamide (Ro
J C Bentley et al.
British journal of pharmacology, 126(7), 1537-1542 (1999-05-14)
1. The present study examined the effects of the selective 5-HT6 receptor antagonist 4-amino-N-(2, 6 bis-methylamino-pyrimidin-4-yl)-benzene sulphonamide (Ro 04-6790) on locomotor activity and unconditioned behaviour in male Sprague Dawley rats (230-300 g). 2. In non-quantified behavioural observations, animals treated with
Bofan Yu et al.
Cell cycle (Georgetown, Tex.), 18(20), 2641-2650 (2019-08-15)
Noncoding RNAs play important roles in the progression of malignant tumors, including triple negative breast cancer (TNBC). Accumulating evidence supported the involvement of the oncogenic MUC1 in tumor metastasis. Our study aimed to explore the roles of miR-140-5p and MUC1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service