Skip to Content
Merck

Skip To

PHG0016

Glycine hydrochloride

Synonym(s):

Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
H2NCH2CO2H · HCl
CAS Number:
Molecular Weight:
111.53
UNSPSC Code:
12352209
NACRES:
NA.21
EC Number:
227-841-8
MDL number:


biological source

synthetic

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, microbial, heavy metals, arsenic and trace impurities; tested

mp

176-180 °C (lit.)

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity; tested

SMILES string

Cl.NCC(O)=O

InChI

1S/C2H5NO2.ClH/c3-1-2(4)5;/h1,3H2,(H,4,5);1H

InChI key

IVLXQGJVBGMLRR-UHFFFAOYSA-N

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of downstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Glycine hydrochloride


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Questions

Reviews

No rating value

Active Filters