Skip to Content
Merck

Skip To

P3417

Dispase from Bacillus polymyxa

powder, ~0.4 unit/mg solid, suitable for cell culture

Synonym(s):

Proteinase, Bacillus polymyxa neutral

Sign In to View Organizational & Contract Pricing.

About This Item

CAS Number:
UNSPSC Code:
12352204
EC Number:
255-914-4
MDL number:
Specific activity:
~0.4 unit/mg solid


form

powder

specific activity

~0.4 unit/mg solid

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

Other Notes

One unit will hydrolyze casein to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 7.5 at 37°C (color by Folin-Ciocalteu reagent).


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Angustias Page et al.
International journal of oral science, 12(1), 1-1 (2020-01-05)
Odontogenic tumours are a heterogeneous group of lesions that develop in the oral cavity region and are characterized by the formation of tumoural structures that differentiate as teeth. Due to the diversity of their histopathological characteristics and clinical behaviour, the



Questions

Reviews

No rating value

Active Filters