Skip to Content
Merck

Skip To

P1380

L-Proline β-naphthylamide hydrochloride

≥99%, suitable for ligand binding assays

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 g
Please contact Customer Service for Availability
CZK 5,470.00

About This Item

Empirical Formula (Hill Notation):
C15H16N2O · HCl
CAS Number:
Molecular Weight:
276.76
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
MDL number:

CZK 5,470.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Proline β-naphthylamide hydrochloride, ≥99%

Quality Segment

assay

≥99%

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

Cl.O=C(Nc1ccc2ccccc2c1)[C@@H]3CCCN3

InChI

1S/C15H16N2O.ClH/c18-15(14-6-3-9-16-14)17-13-8-7-11-4-1-2-5-12(11)10-13;/h1-2,4-5,7-8,10,14,16H,3,6,9H2,(H,17,18);1H/t14-;/m0./s1

InChI key

UKXYCTFKKXKIPY-UQKRIMTDSA-N

Biochem/physiol Actions

L-Proline β-naphthylamide is a substrate used for testing enzyme activity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P2254P5267A3913
form

powder

form

powder

form

powder

form

powder

technique(s)

ligand binding assay: suitable

technique(s)

ligand binding assay: suitable

technique(s)

ligand binding assay: suitable

technique(s)

ligand binding assay: suitable

assay

≥99%

assay

-

assay

≥99% (TLC)

assay

≥98%

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

color

white to off-white

color

white

color

white to yellow

color

white to off-white


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
P1380-1G04061834354635
P1380-100MG04061832417837

Questions

Reviews

No rating value

Active Filters