Skip to Content
Merck

Skip To

P0666

5α-Pregnan-3β-ol-20-one

Synonym(s):

3β-Hydroxy-5α-pregnan-20-one, 3β-Hydroxy-5α-tetrahydroprogesterone, 5α-Dihydropregnenolone, Allopregnanolone, Isopregnanolone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H34O2
CAS Number:
Molecular Weight:
318.49
UNSPSC Code:
41116107
EC Number:
208-224-2
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


SMILES string

CC(=O)C1CCC2C3CC[C@H]4CC(O)CCC4(C)C3CCC12C





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library