Skip to Content
Merck

Skip To

P0047

PD166793 hydrate

≥98% (HPLC)

Synonym(s):

(S)-2-(4′-Bromo-biphenyl-4-sulfonylamino)-3-methyl-butyric acid hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 mg
Please contact Customer Service for Availability
CZK 10,900.00

About This Item

Empirical Formula (Hill Notation):
C17H18BrNO4S · xH2O
CAS Number:
Molecular Weight:
412.30 (anhydrous basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
solid

CZK 10,900.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

storage temp.

2-8°C

SMILES string

O.CC(C)[C@H](NS(=O)(=O)c1ccc(cc1)-c2ccc(Br)cc2)C(O)=O

InChI

1S/C17H18BrNO4S.H2O/c1-11(2)16(17(20)21)19-24(22,23)15-9-5-13(6-10-15)12-3-7-14(18)8-4-12;/h3-11,16,19H,1-2H3,(H,20,21);1H2/t16-;/m0./s1

InChI key

LUKRECOTGXWIDO-NTISSMGPSA-N

Gene Information

Biochem/physiol Actions

PD166793 is a broad spectrum, but class-selective, MMP inhibitor (low nM at MMP-2 & 3 and low μM at MMP-1, 7 & 9).
PD166793 is a broad spectrum, but class-selective, MMP inhibitor (low nM at MMP-2 & 3 and low μM at MMP-1, 7 & 9). Adding PD166793 to the high-fat diet substantially reduced the rise in blood glucose. The improvement of glucose homeostasis in PD166793-treated ZDF rats was a result of the enhancement of β-cell function. It prevents β-cell dysfunction and diabetes in female ZDF rats on a high-fat diet. It has been shown to block left ventricular remodeling and dysfunction in a rat model of heart failure. Preservation of normoglycemia and normal glucose tolerance in compound PD166793-treated animals is accompanied by preservation of the high serum insulin levels that are a consequence of the insulin resistance present in these animals. It is not reversing insulin resistance these animals. The most striking effect of the compound is on pancreatic insulin content and β-cell volume.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A2846G5793SML0178
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

>98% (HPLC)

assay

≥98% (HPLC)

form

solid

form

solid

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

2-8°C

Gene Information

Gene Information

-

Gene Information

human ... HTR1B(3351), HTR1D(3352)

Gene Information

-


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
P0047-25MG04061833213216
P0047-5MG04061833213223

Questions

Reviews

No rating value

Active Filters