Skip to Content
Merck

Skip To

N9663

NEK2, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HsPK21, NLK1

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Form:
buffered aqueous glycerol solution
Assay:
≥70% (SDS-PAGE)
Recombinant:
expressed in baculovirus infected Sf9 cells
Mol wt:
~76 kDa


recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

237-321 nmol/min·mg

mol wt

~76 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NEK2(4751)

Biochem/physiol Actions

NEK2 is closely related in its catalytic domain to the serine/threonine protein kinase NIMA of Aspergillus nidulans that is required for entry into mitosis and may function in parallel to the universal mitotic inducer p34cdc2. Like NIMA, the NEK2 protein is almost undetectable during G1 but accumulated progressively throughout S, reaching maximal levels in late G2.NEK2 is shown to be expressed most abundantly in the testis of the adult tissues examined being localized to the nucleus.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
N9663-10UG04061837906633

Questions

Reviews

No rating value

Active Filters