Skip to Content
Merck

Skip To

N3289

N2-Ethyl-2′-deoxyguanosine

≥98% (HPLC), solid

Synonym(s):

N2-Et-dG

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C12H17N5O4
CAS Number:
Molecular Weight:
295.29
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Quality level:


Quality Level

assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

H2O: >5 mg/mL

storage temp.

2-8°C

SMILES string

CCNC1=Nc2c(ncn2[C@H]3C[C@H](O)[C@@H](CO)O3)C(=O)N1

InChI

1S/C12H17N5O4/c1-2-13-12-15-10-9(11(20)16-12)14-5-17(10)8-3-6(19)7(4-18)21-8/h5-8,18-19H,2-4H2,1H3,(H2,13,15,16,20)/t6-,7+,8+/m0/s1

InChI key

VOKQFDULHQUWAV-XLPZGREQSA-N

General description

N2-Et-dG, an adduct between DNA and the first metabolite of ethanol (acetaldehyde), strongly blocks transcription by RNA polymerases (RNAPs), including mammalian RNAPII and yeast RNAPII, E. coli RNAP, as well as T7 RNAP. All three of the multisubunit RNAPs incorporate a single rNTP residue opposite the N2-Et-dG lesion. The mammalian RNAPII exclusively incorporates cytidine monophosphate (CMP) opposite the N2-Et-dG lesion. The accessory transcription factor TFIIS does not usually act as a lesion bypass factor; instead, it stimulates the polymerase to remove the CMP incorporated opposite the lesion by mammalian RNAPII. N2-Et-dG reduces CTP incorporation by ~1500 fold.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H5653SML1296288104
form

solid

form

-

form

powder

form

solid

assay

≥98% (HPLC)

assay

≥98% (TLC)

assay

≥97% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

solubility

H2O: >5 mg/mL

solubility

-

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

DMSO: 10 mg/mL

color

white to off-white

color

-

color

white to brown

color

white


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Carc. 2 - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Liang Qi et al.
Talanta, 209, 120523-120523 (2020-01-02)
DNA adducts usually occur at a level of 0.1-1 adducts per 108 unmodified DNA bases, which can cause many adverse health effects if not normally repaired. Using microgram amounts of DNA to detect DNA adducts remains challenging due to the
Yuan-Jhe Chang et al.
Archives of toxicology, 92(8), 2665-2680 (2018-06-27)
Adductomics is expected to be useful in the characterization of the exposome, which is a new paradigm for studying the sum of environmental causes of diseases. DNA adductomics is emerging as a powerful method for detecting DNA adducts, but reliable



Global Trade Item Number

SKUGTIN
N3289-25MG04061832632308
N3289-5MG04061832632315

Questions

Reviews

No rating value

Active Filters