Skip to Content
Merck
All Photos(1)

Key Documents

MSST0049

Sigma-Aldrich

SILuProt AKT1,RAC-alpha serine/threonine-protein kinase human

recombinant, expressed in HEK 293 cells, SIL MS Protein Standard, 13C- and 15N-labeled

Synonym(s):

Protein kinase B (PKB), Protein kinase B alpha (PKB alpha), Proto-oncogene c-Akt, RAC-PK-alpha

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CZK 17,400.00

CZK 17,400.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
CZK 17,400.00

About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

CZK 17,400.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in HEK 293 cells

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

potency

≥98% (Heavy amino acids incorporation efficiency by MS)

suitability

suitable for mass spectrometry (standard)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... AKT1 (207)

General description

SILuProt AKT1 is a recombinant, stable isotope-labeled human AKT1 which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in human 293 cells, it is designed to be used as an internal standard for bioanalysis of AKT1 in mass-spectrometry. SILuProt TIMP1 is a protein of 501 amino acids (including N-terminal polyhistidine and flag tags), with a calculated molecular mass of 58.8 kDa.

Sequence

MDYKDDDDKGHHHHHHHHGGQMSDVTIVKEGWVQKRGEYIKNWRPRYFLLKTDGSFIGYKEKPQDVDLPYPLNNFSVAKCQLMKTERPKPNTFIIRCLQWTTVIERTFHVDTPEEREEWTEAIQAVADRLQRQEEERMNCSPTSQIDNIGEEEMDASTTHHKRKTMNDFDYLKLLGKGTFGKVILVREKASGKYYAMKILKKEVIIAKDEVAHTLTESRVLKNTRHPFLTSLKYSFQTKDRLCFVMEYVNGGELFFHLSRERVFSEDRTRFYGAEIVSALDYLHSGKIVYRDLKLENLMLDKDGHIKITDFGLCKEGITDAATMKTFCGTPEYLAPEVLEDNDYGRAVDWWGLGVVMYEMMCGRLPFYNQDHEKLFELILMEDIKFPRTLSSDAKSLLSGLLIKDPNKRLGGGPDDAKEIMRHSFFSGVNWQDVYDKKLVPPFKPQVTSETDTRYFDEEFTAQTITITPPEKYDEDGMDCMDNERRPHFPQFSYSASGRE

Physical form

Supplied as a liquid containing phosphate buffered saline.

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), [email protected].
SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service