Skip to Content
Merck

Skip To

M9904

Worthington Accessories

measuring rod for 35 L Dewar

Synonym(s):

Dewar for liquid nitrogen rod, R033-8C11, cryogenic dewar rid, cryogenic storage rod, measuring rod

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
CZK 513.00

About This Item

NACRES:
NB.22
UNSPSC Code:
41103000
Packaging:
pack of 1 ea

CZK 513.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pack of 1 ea

manufacturer/tradename

Worthington R033-8C11

technique(s)

cryopreservation: suitable

Application

For determination of liquid nitrogen level.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C0671C0796C7431
manufacturer/tradename

Worthington R033-8C11

manufacturer/tradename

Worthington D004-9C11

manufacturer/tradename

Worthington D010-9C16

manufacturer/tradename

Worthington D025-9C16

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

technique(s)

cryopreservation: suitable

technique(s)

-

technique(s)

-

technique(s)

cryopreservation: suitable


Still not finding the right product?

Explore all of our products under Worthington Accessories




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
M9904-1EA04061833452530