Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.56
UNSPSC Code:
23151817
Pricing and availability is not currently available.
Application
Mimetic Yellow 1 is used in affinity and protein chromatography. Mimetic Yellow 1 has been used in studies to develop a potential application in regulating fertility of the ram.
1 of 1
This Item | |||
|---|---|---|---|
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in dry ice |
| description 10 ml | description - | description 10 ml | description - |
| form suspension | form suspension | form suspension | form suspension |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
signalword
Warning
hcodes
Hazard Classifications
Flam. Liq. 3
Storage Class
3 - Flammable liquids
wgk
WGK 1
flash_point_f
95.0 °F - closed cup
flash_point_c
35 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




