Skip to Content
Merck

Skip To

M6909

D-Mannoheptose

≥99%

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C7H14O7
CAS Number:
Molecular Weight:
210.18
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:


assay

≥99%

form

powder

optical activity

[α]20/D 69.0 to 73.0 °, c = 4% (w/v) in water

technique(s)

gas chromatography (GC): suitable

SMILES string

OCC(O)C(O)C(O)C(O)C(O)C=O

InChI

1S/C7H14O7/c8-1-3(10)5(12)7(14)6(13)4(11)2-9/h1,3-7,9-14H,2H2

InChI key

YPZMPEPLWKRVLD-UHFFFAOYSA-N

General description

D-Mannoheptose is an aldoheptose. [1]

Application

D-Mannoheptose has been used in a study to assess the lipopolysaccharide biosynthesis of Shigella sonnei. [2]

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


P Courtois et al.
International journal of molecular medicine, 6(2), 171-175 (2000-07-13)
The possible use of D-mannoheptose or D-glycero-D-gulo-heptose as substitute of D-mannoheptulose for specific inhibition of D-glucose phosphorylation, metabolism and insulinotropic action was investigated in the present study. The two aldoheptoses failed to duplicate the effect of D-mannoheptulose upon the phosphorylation
T Kontrohr et al.
The Journal of biological chemistry, 256(15), 7715-7718 (1981-08-10)
From a Shigella sonnei R mutant which incorporates into its cell wall lipopolysaccharide D-glycero-D-mannoheptose and contains no L-glycero-D-mannoheptose, a nucleotide-linked sugar was isolated and identified as adenosine 5'-diphosphate-D-glycero-d-mannoheptose by chemical and chromatographic analysis. This intermediary compound is assumed to play



Global Trade Item Number

SKUGTIN
M6909-1G04061826060827

Questions

Reviews

No rating value

Active Filters