Skip to Content
Merck
All Photos(1)

Key Documents

M5566

Sigma-Aldrich

Anti-Myosin IX/Myr5 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-CELIAC4, Anti-MYR5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 230 kDa

species reactivity

rat

technique(s)

microarray: suitable
western blot: 1:1,000 using rat embryonic brain extract.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Myosin IX belongs to the family of unconventional myosins. The tail region of these myosins contains a GTPase-activating protein (GAP) domain. It is expressed at higher levels in the embryonic brain than in the adult brain. In the adult rat, it is highly expressed in the lung, liver, spleen, and testis. The subcellular localization of class IX myosins appears to be partly cytoplasmic and partly associated with membranes and the actin cytoskeleton. The tail domain of myosin IX/Myr5 contains a region that coordinates two atoms of zinc. Myosin IX is thought to be involved in signal transduction and leukocyte differentiation.
Myosin IX/Myr5 is a single headed myosin which possesses an N-terminal extension and a large loop 2 insertion.

Specificity

Anti-Myosin IX/Myr5 recognizes rat myosin IX/Myr5 (230 kDa).

Immunogen

synthetic peptide corresponding to the C-terminal region of rat myosin IX/Myr5 (amino acids 1960-1980).

Application

Anti-Myosin IX/Myr5 antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Myosin IX expression level may vary during development and differentiation. The tail domain of myosin IX/Myr5 and the GTPase-activating protein (GAP) domain stimulates GTP hydrolysis. It is implicated in the regulation of actin organization, and the actin cytoskeleton. Overexpression of both Myr5 and Myr7 in cultured cells results in inactivation of Rho, loss of actin stress fibers, and focal contacts, leading to changes in cell morphology.
Myosin IX/Myr5 moves along the actin filaments for long distances and is mostly present in its ATP-bound state. It has been studied as a negative regulator of RhoC which is a G-protein.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For prolonged storage, freeze in working aliquots at −20 °C. Repeated freezing and thawing is not recom-mended. Storage in frost-free freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wanqin Liao et al.
The Journal of biological chemistry, 285(32), 24933-24942 (2010-06-12)
Mammalian myosin IXb (Myo9b) has been shown to exhibit unique motor properties in that it is a single-headed processive motor and the rate-limiting step in its chemical cycle is ATP hydrolysis. Furthermore, it has been reported to move toward the
Vijayalaxmi Nalavadi et al.
The Journal of biological chemistry, 280(47), 38957-38968 (2005-09-24)
Myosin IXb (Myo9b) was reported to be a single-headed, processive myosin. In its head domain it contains an N-terminal extension and a large loop 2 insertion that are specific for class IX myosins. We characterized the kinetic properties of purified
Sandra Struchholz et al.
The Journal of biological chemistry, 284(6), 3663-3671 (2008-12-09)
The mammalian class IX myosin Myo9b can move considerable distances along actin filaments before it dissociates. This is remarkable, because it is single headed and because the rate-limiting step in its ATPase cycle is ATP hydrolysis. Thus, it spends most
Arturo Raya-Sandino et al.
Biochimica et biophysica acta. Molecular cell research, 1864(10), 1714-1733 (2017-05-31)
Silencing Zonula occludens 2 (ZO-2), a tight junctions (TJ) scaffold protein, in epithelial cells (MDCK ZO-2 KD) triggers: 1) Decreased cell to substratum attachment, accompanied by reduced expression of claudin-7 and integrin β1, and increased vinculin recruitment to focal adhesions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service