Skip to Content
Merck
All Photos(1)

Key Documents

M5063

Sigma-Aldrich

Myelin-Associated Glycoprotein/Fc Chimera from rat

>95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

MAG

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 9,950.00

CZK 9,950.00

List PriceCZK 19,900.00Save 50%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
CZK 9,950.00

About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.83

CZK 9,950.00

List PriceCZK 19,900.00Save 50%

Please contact Customer Service for Availability

Request a Bulk Order

biological source

rat

Quality Level

recombinant

expressed in NSO cells

Assay

>95% (SDS-PAGE)

form

lyophilized powder

mol wt

120 kDa by SDS-PAGE
calculated mol wt ~81 kDa

impurities

endotoxin, tested

UniProt accession no.

application(s)

cell analysis

storage temp.

−20°C

Gene Information

rat ... Mag(29409)

Related Categories

Biochem/physiol Actions

MAG is a type I transmembrane glycoprotein containing five Ig-like domains in its extracellular domain. It is an adhesion molecule belonging to the immunoglobin superfamily. These adhesion molecules bind specifically to cell-surface glycan containing sialic acid residues that define the I-type sialyl lectin subgroup. Thus, they are also called the sialoadhesin family. Sialoadhesins mediate diverse biological processes through recognition of specific sialyted glycans on the cell surface. MAG, a minor component of myelin in the central and peripheral nervous system, has been implicated in the formation and maintenance of myelin. MAG is expressed on myelinating oligodenrocytes and Schwann cells, and preferentially recognize a 2,3-linked sialic acid on O-linked glycans and gangliosides. MAG exists as two isoforms that differ in the sequence and length of the cytoplasmic tail. The large isoform (71 kDa) and small isoform (67 kDa) arise from alternative splicing of mRNAs. Lymphocytes under pathologic conditions, it would normally interact with neuronal cells. It has been shown that MAG promotes axonal growth from neonatal dorsal root ganglion (DRG) neurons and embryonic spinal neurons, but is a potent inhibitor of axonal re-growth from adult DRG and postnatal cerebellar neurons. MAG plays an important role in the interaction between axons and myelin. A soluble form of MAG containing the extracellular domain is released from myelin in large quantities and identified in normal human tissues and in tissues from patients with neurological disorders. This soluble MAG may contribute to the lack of neuronal regeneration after injury.
MAG, a member of the immunoglobin superfamily, is expressed on myelinating oligodendrocytes and Schwann cells. MAG plays an important role in the interacation between axons and myelin.

Other Notes

Extracellular domain of rat myelin-associated glycoprotein (MAG) fused to the C-terminal Fc region of human IgG1.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Analysis Note

Measured by its ability to inhibit neurite outgrowth of cultured embryonic chick dorsal root ganglia neurons.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kelm et al.
Current biology : CB, 4(11), 965-972 (1994-11-01)
Protein-carbohydrate interactions are believed to be important in many biological processes that involve cell-cell communication. Apart from the selectins, the only well-characterized vertebrate sialic acid-dependent adhesion molecules are CD22 and sialoadhesin; CD22 is a member of the immunoglobulin superfamily that
J L Salzer et al.
The Journal of cell biology, 104(4), 957-965 (1987-04-01)
The myelin associated glycoproteins (MAG) are integral plasma membrane proteins which are found in oligodendrocytes and Schwann cells and are believed to mediate the axonal-glial interactions of myelination. In this paper we demonstrate the existence in central nervous system myelin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service