Merck
Skip To
M3782
Metabolic Pathways, 22nd Ed.
Chart, 12.5 × 19 in.
Metabolic Pathways, 22nd Ed.
Sign In to View Organizational & Contract Pricing.
About This Item
NACRES:
NA.28
UNSPSC Code:
41100000
publication info
D.E. Nicholson, Sigma Chemical Co., St. Louis, MO, 2003
General description
This 22nd edition of the IUBMB-Nicholson Metabolic Pathways Chart features an enhanced ATP metabolism in mitochondria and chloroplasts. It illustrates the sequences of reactions involved in different aspects of metabolism and how they are integrated. The backbone of the map is the Glycolytic Pathway, followed by the TCA (Krebs) Cycle and the ElectronTransport System, which together produce most of the energy (ATP) required for all life. Most other aspects of metabolism are associated with this backbone and are differentiated by the use of color. Human metabolism is distinguished by the use of black arrows. Some 550 reactions are identified by their IUBMB Enzyme Commission (EC) numbers which are then indexed.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
I8014M390715-107
publication info
D.E. Nicholson, Sigma Chemical Co., St. Louis, MO, 2003
publication info
D.E. Nicholson, Sigma Chemical Co., St. Louis, MO, 1998
publication info
D.E. Nicholson, Sigma Chemical Co., St. Louis, MO, 2003
publication info
-
Still not finding the right product?
Explore all of our products under Metabolic Pathways, 22nd Ed.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Active Filters
  1. USA
    • Review 1
    • Votes 0
    1 out of 5 stars.
    Print too small to be useful
    The Metabolic pathways poster I received is 12 x 19 but the letters are so small as to make it unusable--at least for me. I love the concept and the clean look of the product but it is impractical and not useful at all. I ordered the larger sized poster as well but it has been on backorder for more than a year so I haven't received it yet. I'm very disappointed in the product.
    Helpful?