Skip to Content
Merck

Skip To

M150

S(+)-Methadone hydrochloride

solid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H27NO · HCl
CAS Number:
Molecular Weight:
345.91
UNSPSC Code:
12352200
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

color

white

solubility

H2O: soluble

Other Notes

Enantiomer of methadone that is either inactive or a weak agonist at opioid receptors; NMDA antagonist that blocks the development of morphine tolerance.


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Bath Salts in Urine - Supel-Select SCX SPE and Ascentis Express HILIC Application Note


A M Davis et al.
The Journal of pharmacology and experimental therapeutics, 289(2), 1048-1053 (1999-04-24)
Previous in vitro and in vivo studies have determined that the d isomer of methadone has N-methyl-D-aspartate (NMDA) receptor antagonist activity. The present studies examined the ability of d-methadone to attenuate the development of morphine tolerance in mice and rats
A L Gorman et al.
Neuroscience letters, 223(1), 5-8 (1997-02-14)
Racemic (dl)-methadone has antagonist activity at the N-methyl-D-aspartate (NMDA) receptor. We evaluated dl-methadone, the opioid active (l-) and the opioid inactive (d-) isomers in competition binding assays. dl-Methadone and its d- and l- isomers exhibited low micromolar affinities for the
K Kristensen et al.
Life sciences, 56(2), PL45-PL50 (1995-01-01)
The binding affinities of racemic methadone and its optical isomers R-methadone and S-methadone were evaluated for the opioid receptors mu1, mu2, delta and kappa, in comparison with that of morphine. The analgesic R-methadone had a 10-fold higher affinity for mu1