Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Linear Formula:
CH2=C(CH3)COOCH3
CAS Number:
Molecular Weight:
100.12
EC Number:
201-297-1
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
605459
Pricing and availability is not currently available.
vapor density
3.5 (vs air)
vapor pressure
29 mmHg ( 20 °C)
assay
≥99% (GC)
autoignition temp.
815 °F
contains
10 ppm hydroquinone monomethyl ether as inhibitor
expl. lim.
12.5 %
refractive index
n20/D 1.414 (lit.)
bp
100 °C (lit.)
mp
−48 °C (lit.)
density
0.936 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
COC(=O)C(C)=C
InChI
1S/C5H8O2/c1-4(2)5(6)7-3/h1H2,2-3H3
InChI key
VVQNEPGJFQJSBK-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under Methyl methacrylate
signalword
Danger
hcodes
Hazard Classifications
Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk
WGK 1
flash_point_f
50.0 °F - closed cup
flash_point_c
10 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

