Skip to Content
Merck

Skip To

L7387

Lanolin

Synonym(s):

Wool Grease, Wool Wax, Wool fat

Sign In to View Organizational & Contract Pricing.

About This Item

CAS Number:
UNSPSC Code:
12352211
EC Number:
232-348-6
NACRES:
NA.25
MDL number:


autoignition temp.

833 °F

lipid type

oils

Application

Lanolin has been used as a constituent of VALAP (Vaseline, lanolin, paraffin) to stabilize glass slides during worm-immobilization method. It has also been used to transport auxin in to Arabidopsis to induce petiole torsions.

Biochem/physiol Actions

Lanolin is a complex waxy substance containing varying quantities of long-chain waxy esters and lanolin alcohols, acids and polycarbons. It is isolated from wool of sheep. Lanolin is used in a wide variety of hydrophobic barrier creams, ointments and emollients. It may also be used as a component of lubricants, rust-preventative coatings, shoe polish and other commercial products. lanolin is often studied to determine its allergenic properties.
Lanolin is a complex waxy substance composed of varying quantities of long-chain waxy esters and lanolin alcohols, acids and polycarbons. Lanolin is used in a wide variety of creams, ointments and emollients. Lanolin is often studied to determine it allergenic properties.


Still not finding the right product?

Explore all of our products under Lanolin


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters