Skip to Content
Merck

Skip To

L6016

Lipopolysaccharides (rough strains) from Salmonella enterica serotype typhimurium TV119 (Ra mutant)

Synonym(s):

LPS

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:


biological source

Salmonella enterica (Serotype typhimurium TV119 Ra mutant)

form

lyophilized powder

impurities

<3% Protein (Lowry)

shipped in

ambient

storage temp.

2-8°C

General description

This product is phenol:chloroform:petroleum ether extracted from Salmonella typhimurium strain TV119(Ra mutant). The source strain is from a private collection.

Biochem/physiol Actions

Lipopolysaccharides (LPS) are localized in the outer layer of the membrane and are, in noncapsulated strains, exposed on the cell surface. They contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics.

Preparation Note

Prepared by phenol-chloroform-petroleum ether extraction.

The product is soluble in water (5 mg/ml) or cell culture medium (1 mg/ml) yielding a hazy, faint yellow solution. A more concentrated, though still hazy, solution (20 mg/ml) has been achieved in aqueous saline after vortexing and warming to 70-80 oC. Lipopolysaccharides are molecules that form micelles in every solvent. Hazy solutions are observed in water and phosphate buffered saline. Organic solvents do not give clearer solutions. Methanol yields a turbid suspension with floaters, while water yields a homogeneously hazy solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
L6016-50MG04061833275030
L6016-10MG04061833275023

Questions

Reviews

No rating value

Active Filters