Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C54H84N12O18
CAS Number:
Molecular Weight:
1189.31
UNSPSC Code:
12352200
MDL number:
Pricing and availability is not currently available.
assay
≥95% (HPLC)
storage temp.
−20°C
SMILES string
CC(O)C(NC(=O)C(CCC(O)=O)NC(=O)C1CCCN1C(=O)C(CCC(O)=O)NC(=O)C2CCCN2C(=O)C3CCCN3C(=O)C4CCCN4C(=O)C(NC(=O)C5CCCN5C(=O)C6CCCN6C(=O)C(N)CCCCN)C(C)O)C(O)=O
Other Notes
Carboxy-terminal sequence of SV40 tumor antigen
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.