Skip to Content
Merck

Skip To

I9909

Monoclonal Anti-Insulin-like Growth Factor-I antibody produced in mouse

clone 56408, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-IGF1

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
51111800
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
56408, monoclonal
Application:
ELISA (c), WB
Citations:
3


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

56408, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

capture ELISA: 2-8 μg/mL, western blot: 1-2 μg/mL

isotype

IgG1

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IGF1(3479)

Immunogen

purified, E. coli-derived recombinant human insulin-like growth factor-I.

Biochem/physiol Actions

Insulin-like growth factor -1 shares structural homology with proinsulin and is mainly produced by liver. IGF-1 acts via specific receptor, IGF-1R, a heterodimeric tyrosine kinase receptor that signals to various second messenger pathways such as MAPK, Raf/Ras and PI3K cascades. The effects of IGF-1R signaling are important at many developmental levels from cells to organ to organism, growth, differentiation, anti-apoptosis, proliferation and transformation.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions






Global Trade Item Number

SKUGTIN
I9909-500UG04061832714899

Questions

Reviews

No rating value

Active Filters