Skip to Content
Merck
All Photos(6)

Documents

HPA017960

Sigma-Aldrich

Anti-CORO2B antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Clipin-C, Anti-Coronin-2B, Anti-Coronin-like protein C, Anti-Protein FC96

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

YQEDIYPMTPGTEPALTPDEWLGGINRDPVLMSLKEGYKKSSKMVFKAPIKEKKSVVVNGIDLLENVPPRTENELLRMFFRQQDEIRRLKEELAQKDIRIRQLQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CORO2B(10391)

General description

Coronin actin binding protein, 2B (CORO2B) belongs to the coronin-like protein (Clipin) family and is a type II coronin isoform. It is expressed in the brain, and possesses a WD domain and an α-helical region. The gene encoding CORO2B is located on human chromosome 15.

Immunogen

Coronin-2B recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Coronin actin binding protein, 2B (CORO2B) is a component of the cross-bridge between the plasma membrane and actin cytoskeleton. CORO2B may also function in the rearrangement of neuronal actin.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72566

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keefe T Chan et al.
Trends in cell biology, 21(8), 481-488 (2011-06-03)
Coronins are a conserved family of actin cytoskeleton regulators that promote cell motility and modulate other actin-dependent processes. Although these proteins have been known for 20 years, substantial progress has been made in the past 5 years towards their understanding.
Yuewen Chen et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 43(2), 211-220 (2023-01-15)
In the developing cortex, excitatory neurons migrate along the radial fibers to their final destinations and build up synaptic connection with each other to form functional circuitry. The shaping of neuronal morphologies by actin cytoskeleton dynamics is crucial for neuronal
Z M Ahmed et al.
Clinical genetics, 75(1), 86-91 (2008-05-29)
Usher syndrome (USH) is a hereditary disorder associated with sensorineural hearing impairment, progressive loss of vision attributable to retinitis pigmentosa (RP) and variable vestibular function. Three clinical types have been described with type I (USH1) being the most severe. To
Hongjiao Wu et al.
Cell death & disease, 15(6), 457-457 (2024-06-28)
In eukaryotes, the nucleolus is the critical non-membranous organelle within nuclei that is responsible for ribosomal DNA (rDNA) transcription and ribosome biogenesis. The transcription of rDNA, a rate-limiting step for ribosome biogenesis, is tightly regulated to meet the demand for
T Nakamura et al.
The Journal of biological chemistry, 274(19), 13322-13327 (1999-05-01)
Brain-enriched human FC96 protein shows a close sequence similarity to the Dictyostelium actin-binding protein coronin, which has been implicated in cell motility, cytokinesis, and phagocytosis. A phylogenetic tree analysis revealed that FC96 and two other mammalian molecules (p57 and IR10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service