Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
10 μg | Please contact Customer Service for Availability | CZK 6,440.00 |
About This Item
CZK 6,440.00
Product Name
Interleukin-17F human, HumanKine®, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin, tested
biological source
human
Quality Segment
recombinant
expressed in HEK 293 cells
assay
≥95% (SDS-PAGE)
form
lyophilized powder
potency
≤10 ng/mL EC50
mol wt
dimer 38 kDa (glycosylated)
packaging
pkg of 10 μg
storage condition
avoid repeated freeze/thaw cycles
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... IL17F(112744)
Physical form
Analysis Note
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| biological source human | biological source human | biological source human | biological source human |
| recombinant expressed in HEK 293 cells | recombinant expressed in HEK 293 cells | recombinant expressed in HEK 293 cells | recombinant expressed in HEK 293 cells |
| assay ≥95% (SDS-PAGE) | assay ≥95% (SDS-PAGE) | assay ≥95% (SDS-PAGE) | assay >95% (SDS-PAGE) |
| technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
Still not finding the right product?
Explore all of our products under Interleukin-17F human
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



