Skip to Content
Merck

Skip To

H6029

7-Hydroxycoumarin β-D-glucuronide sodium salt

≥95% (HPLC)

Synonym(s):

Umbelliferyl β-D-glucuronide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 mg
Please contact Customer Service for Availability
CZK 8,360.00

About This Item

Empirical Formula (Hill Notation):
C15H13NaO9
CAS Number:
Molecular Weight:
360.25
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:

CZK 8,360.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

solid

impurities

5% water (Karl Fischer)

color

white

storage temp.

2-8°C

SMILES string

[Na+].O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C([O-])=O)Oc2ccc3C=CC(=O)Oc3c2

InChI

1S/C15H14O9.Na/c16-9-4-2-6-1-3-7(5-8(6)23-9)22-15-12(19)10(17)11(18)13(24-15)14(20)21;/h1-5,10-13,15,17-19H,(H,20,21);/q;+1/p-1/t10-,11-,12+,13-,15+;/m0./s1

InChI key

SKHLGBDGEPDEME-KSOKONAESA-M

Biochem/physiol Actions

Metabolite of 7-ethoxycoumarin and coumarin. Useful as a pharmacokinetic standard in in vitro metabolic studies.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
R288643073SMB00948
assay

≥95% (HPLC)

assay

≥90%

assay

≥98.5% (HPLC)

assay

≥95% (HPLC)

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

200

form

solid

form

powder

form

-

form

solid

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

color

white

color

-

color

-

color

white to beige

impurities

5% water (Karl Fischer)

impurities

-

impurities

≤12.0% water

impurities

-


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
H6029-1MG04061826059760
H6029-5MG04061832391250

Questions

Reviews

No rating value

Active Filters