Skip to Content
Merck
All Photos(1)

Key Documents

EP6336000040

Eppendorf® Mastercycler® Nexus X2 Thermal Cyclers

Nexus GX2, complete unit, aluminum block, AC/DC input 230 V, 50/60 Hz (GB)

Synonym(s):

Eppendorf Thermocycler, Thermal Cycler

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 304,700.00

CZK 304,700.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
CZK 304,700.00

About This Item

UNSPSC Code:
41102404
NACRES:
NC.21

CZK 304,700.00


Please contact Customer Service for Availability

feature

CSA and CE compliant
Nexus GX2
aluminum block
complete unit
holds 64/32?×?0.2?mL PCR tubes or 24/10?×?0.5 mL PCR tubes
unit can control 2 other units in network

manufacturer/tradename

Eppendorf® 6336000040 (GB)

parameter

1-12 °C (gradient range)
30-99 °C temp. range (gradient)
37-110 °C temp. range (Lid)
4-99 °C temp. range (control)

technique(s)

PCR: suitable

W × D × H

25 cm × 41.2 cm × 32.1 cm

weight

11 kg

Looking for similar products? Visit Product Comparison Guide

General description

The Mastercycler nexus X2 gives you the ability to run two totally independent protocols at the same time. Smaller assays fit nicely on the 32-well-block – larger assays can exceed 48 samples and run on the 64-well-block. The larger block is available with a gradient function. As every member of the Mastercycler nexus family, the Mastercycler nexus X2 can be combined with all other family members in a network of up to 3 units. In combination with Eppendorf′s PCR Tubes, PCR Strips or divisible plates, the Mastercycler nexus X2 will give you consistent and publishable results every day!
Certified Quality
All Eppendorf cyclers follow these quality guidelines:
  • Individual, documented quality control certificates
  • Calibration accordingly to national and international standards: NIST (USA), DKD/PTB (Germany), UKAS/NPL (UK)
  • UL/cUL listed

Application

Standard PCR, Fast PCR, PCR optimization

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Mastercycler is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service