EP0030120175
Eppendorf® Safe-Lock micro test tubes
capacity 1.5 mL, blue, pack of 1000 ea
Synonym(s):
Eppendorf tube, Eppendorfs
Select a Size
CZK 1,940.00
Select a Size
About This Item
CZK 1,940.00
Recommended Products
material
blue
cap (push fit)
conical bottom
polypropylene
sterility
non-sterile
packaging
pack of 1000 ea
manufacturer/tradename
Eppendorf® 0030120175
capacity
1.5 mL
maximum volume
1.5 mL
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- The hinged Eppendorf® Safe-Lock lid provides additional protection against unintentional lid opening during incubation and storage.
- g-Safe®:Provides great centrifugation stability and prevents sample loss due to tube breakage
- The optimized sealing properties ensures safety when working with hazardous samples and minimal evaporation rates during long-term storage.
- The large frosted lid and wide side-surface area make labeling convenient.
- Functions between temperatures -86 °C to 100 °C
- Autoclavable when open (121 °C, 20 min)
- Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage g-Safe®: Depending on Safe-Lock Tube variant (see manual) exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
- Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
- Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes * Precise lid sealing for minimal evaporation rates during long-term storage
- Large frosted lid and surface on the side for easy labeling
- Ensured functionality from -86 °C to 100 °C
- Autoclavable when open (121 °C, 20 min)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service