EP0030113489
Eppendorf® Conical Tubes with SafeCode system (data matrix code and plain text labels)
non-sterile, screw cap, capacity 15 mL, PCR clean, pack of 10 × 50 ea
Synonym(s):
Conical Centrifuge tube, micro test tubes
About This Item
Recommended Products
material
colorless
conical bottom
screw top cap
sterility
non-sterile
feature
DNase free
PCR clean
neck thread external
packaging
pack of 10 × 50 ea
manufacturer/tradename
Eppendorf® 0030113489
capacity
15 mL
suitability
suitable for PCR
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Features and Benefits
ECC 200 error correction function enables safe code reading with up to 25 % damaged codes
Temperature range: -86 °C to +121 °C
Manufactured from high-quality polypropylene for highest sample purity
Off-the-shelf availability for comfortable handling
ID-specific tube information like lot numbers digitally available via (URL: https://www.eppendorf.com/safecode-data)[Eppendorf dataport] and website for comprehensive documentation
Direct import of codes and all data into (URL: https://www.elabnext.com/eppendorf)[eLabInventory] and (URL: https://www.elabnext.com/eppendorf)[eLabJournal](URL: null)(URL: https://www.elabnext.com/eppendorf)[ software] for convenient handling
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service