Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Please contact Customer Service for Availability | CZK 5,620.00 |
50 μg | Please contact Customer Service for Availability | CZK 10,100.00 |
About This Item
CZK 5,620.00
description
Powered by Eupheria Biotech
Quality Segment
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
CGATGGAGCACCCATATACCACTGGATGAAGAACGGGATCCCCTTCTGCACAGTTTCAGTAACCTGAAAGAGGTCTTAAACAGTGTGACAGAACTCTCAGAGATTGAGCCAAATGTATTCCTTCGTCCATTTCTGGAAGTCATTCGCTCTGAAGACACCACTGGTCCTATCACCGGCCTGGCACTCACCTCTGTCAACAAGTTCCTGTCCTACGCACTCATAGATCCAACTCATGAGGGCACAGCAGAGGGCATGGAGAACATGGCAGATGCTGTCACTCACGCCCGTTTTGTGGGCACAGATCCTGCCAGTGATGAAGTTGTCCTGATGAAAATCCTTCAGGTTCTCCGAACTCTGTTGCTAACCCCAGTGGGTACCCACTTAACAAATGAATCTGTATGTGAGATTATGCAGTCTTGCTTCCGGATTTGCTTTGAAATGAGACTCAGTGAGTTATTGAGAAAATCCGCAGAGCACACTCT
Ensembl | mouse accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
mouse ... GBF1(107338), Gbf1(107338)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| Gene Information mouse ... GBF1(107338), Gbf1(107338) | Gene Information mouse ... NSUN3(106338), Nsun3(106338) | Gene Information mouse ... GUF1(231279), Guf1(231279) | Gene Information mouse ... GBX1(231044), Gbx1(231044) |
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| Ensembl | mouse accession no. | Ensembl | mouse accession no. | Ensembl | mouse accession no. | Ensembl | mouse accession no. |
| esiRNA cDNA target sequence CGATGGAGCACCCATATACCACTGGATGAAGAACGGGATCCCCTTCTGCACAGTTTCAGTAACCTGAAAGAGGTCTTAAACAGTGTGACAGAACTCTCAGAGATTGAGCCAAATGTATTCCTTCGTCCATTTCTGGAAGTCATTCGCTCTGAAGACACCACTGGTCCTATCACCGGCCTGGCACTCACCTCTGTCAACAAGTTCCTGTCCTACGCACTCATAGATCCAACTCATGAGGGCACAGCAGAGGGCATGGAGAACATGGCAGATGCTGTCACTCACGCCCGTTTTGTGGGCACAGATCCTGCCAGTGATGAAGTTGTCCTGATGAAAATCCTTCAGGTTCTCCGAACTCTGTTGCTAACCCCAGTGGGTACCCACTTAACAAATGAATCTGTATGTGAGATTATGCAGTCTTGCTTCCGGATTTGCTTTGAAATGAGACTCAGTGAGTTATTGAGAAAATCCGCAGAGCACACTCT | esiRNA cDNA target sequence ATCCGTAGCTCTGCTGCAATGTGCTTATCCAGGTTATCTTCTCTGTAACGAGTATGATCGCCCGAGAGGAAGATGGCTGAGACAGACGTTGGAATCTTTCATCCCACAGCCTTTGATAAATGTAATTAAAGTGTCTGAATTGGATGGCAGAGAAATGGGAGATGCCCAGCCTGCAACATTTGACAAGGTGTTAGTTGACGCTCCATGTTCAAATGATCGAAGCTGGCTGTTCTCGTCGGACTCTCAGAAAGCTGCCTATAGGATCCATCAAAGGAAGAATTTGCCTGTCCTACAGGTAGAACTCGTAAGGTCTGCAATTAAAGCCTTGCGCCCCGGAGGGCTCCTCGTGTACTCTACATGCACGCTTTCCAAGGCTGAAAATCAAGATGTGATCAGCGAGG | esiRNA cDNA target sequence CAAGTAGAACGAGAAAGAGGAATCACTGTGAAGGCGCAGACAGCCTCTCTGTTTTACAGCTTTGGAGGGAAACAGTACCTTTTAAACCTCATCGACACACCAGGCCACGTTGATTTTAGTTATGAAGTGTCCAGGTCACTGTCTGCATGCCAAGGTGTTTTACTGGTAGTGGATGCAAATGAGGGTATTCAAGCTCAAACTGTGGCCAACTTCTTCCTTGCTTTTGAAGCACAGCTGTCAGTAATTCCAGTTATCAATAAGATAGATCTGAAGAATGCTGACCCTGAAAGGGTGGGAAAGCAGATTGAAAAAGTATTTGATATCCCTAGTGAGGAGTGTATTAAGATTTCTGCGAAACTTGGGACAAACGTTGACAGTGTGCTTCAGGCAGTCATTGAACGAATACCTCCTCCAAAAGTCCATCGTGAAAACCCACTGAAAGCTTTGGTATTTGACTCCACCTTTGACCAGTACAGAGGGGTGATAGCCAATATAGCACTGTTTGATGGAGTGG | esiRNA cDNA target sequence GGCCATAGTTCTTTATTTATTTTGTGTAAAGTTTGTATATATGGAACTATTTTGTCTTTGTCTGTCTGTATCCCAAAAGGGGACTGGGAAATTGTTTCTTTGAACGTTACCCACTTTGTCCAGCCCTACTGCTTTGCTGCTTTAGCACAAGGACATGCAAGGGCATGAAATGGAACTGTCATGGCCAATATGGGGGTCTTCAGCAGCTCCCTCGTCACTCCAGTGTATTCCCACACCTTCTGTTCCCATTGGGCTACTGTATCCCCACTTCCTGTGAACAAACCCATCTGCTTCTGATGTCTTCTCTTTGGGAGAAGCAGCACACTTCAGGAAGAGACATACTTCAGTTTGCACTGAAGGGAGACCCCCCCAGTTTAGCTTGGAGAACGTTCAAGTTATCTACATAGAAGATGCTGTCCGGTCAGTGATTAAGGCAGGGCAGGAAGCGTACTCCACCACCTGAGCTGAACAGAGAGGGGATTCCTTTACAGGGGCACACAGCCTCATTTCTGT |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
