Skip to Content
Merck
All Photos(1)

Key Documents

E7773

Sigma-Aldrich

pFLAG-CMV-5c Expression Vector

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CZK 15,900.00

CZK 15,900.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
CZK 15,900.00

About This Item

UNSPSC Code:
41106610

CZK 15,900.00


Please contact Customer Service for Availability

storage temp.

−20°C

Quality Level

Application

For cytoplasmic expression

Legal Information

pFLAG-CMV is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service