Skip to Content
Merck
All Photos(1)

Key Documents

E5007

Supelco

Etonitazene hydrochloride

Synonym(s):

1-(Diethylaminoethyl)-2-(p-ethoxybenzyl)-5-nitrobenzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28N4O3 · HCl
CAS Number:
Molecular Weight:
432.94
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

drug control

USDEA Schedule I; Home Office Schedule 1; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

Cl.CCOc1ccc(Cc2nc3cc(ccc3n2CCN(CC)CC)[N+]([O-])=O)cc1

InChI

1S/C22H28N4O3.ClH/c1-4-24(5-2)13-14-25-21-12-9-18(26(27)28)16-20(21)23-22(25)15-17-7-10-19(11-8-17)29-6-3;/h7-12,16H,4-6,13-15H2,1-3H3;1H

InChI key

JTBRYBUIGVMMQL-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

A highly selective μ opioid receptor agonist. Reported to be 100-1000 times more potent than morphine in receptor binding studies and in behavioral studies.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T May et al.
Neuroscience letters, 275(2), 109-112 (1999-11-24)
Rats, for 8 weeks consuming the mu-opioid agonist etonitazene (forced and free choice conditions yielding high and low drug-consumers), were sacrificed after 2 days or 6 weeks lasting drug deprivation. Binding characteristics of membranes from the parieto-occipital cortex of these
Lionel Moulédous et al.
FEBS letters, 589(18), 2401-2408 (2015-08-01)
Many in vitro data have shown that the efficacy of several opioid drugs is correlated with differential mu-opioid (MOP) receptor phosphorylation. Label-free semiquantitative on-line nanoflow liquid chromatography-tandem mass spectrometry (nanoLC-MS/MS) analyses were performed to compare the endogenous MOP receptor phosphorylation
T P Piepponen et al.
Pharmacology, biochemistry, and behavior, 63(2), 245-252 (1999-06-17)
The role of mu1-opioid receptors in the acceleration of cerebral dopaminergic transmission induced by morphine and the putative mu1-opioid agonist, etonitazene, was studied in rats by measuring the tissue levels of dopamine (DA) and its metabolites 3,4-dihydroxyphenylacetic acid (DOPAC) and
G Zernig et al.
Life sciences, 57(23), 2113-2125 (1995-01-01)
In order to quantitate the extent to which opioid agonist potencies obtained in behavioral assays are determined by the apparent in vivo affinity and efficacy of the agonist, the antinociceptive effects of the mu opioid agonists morphine, fentanyl, etonitazene, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service