Skip to Content
Merck

Skip To

D6250

2′-Deoxyadenosine 5′-monophosphate sodium salt

Sigma Grade, ≥96% (HPLC)

Synonym(s):

2′-Deoxyadenylic acid, dAMP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H14N5O6P · xNa+
CAS Number:
Molecular Weight:
331.22 (free acid basis)
UNSPSC Code:
41106305
PubChem Substance ID:
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Sigma Grade

assay

≥96% (HPLC), ≥96% (spectrophotometric assay)

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Na+].[Na+].Nc1ncnc2n(cnc12)[C@H]3C[C@H](O)[C@@H](COP([O-])([O-])=O)O3

InChI

1S/C10H14N5O6P.2Na/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(16)6(21-7)2-20-22(17,18)19;;/h3-7,16H,1-2H2,(H2,11,12,13)(H2,17,18,19);;/q;2*+1/p-2/t5-,6+,7+;;/m0../s1

InChI key

SNYHHRDGEJPLGT-OJSHLMAWSA-L



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Sachie Shibayama et al.
Journal of chromatography. A, 1406, 210-214 (2015-07-05)
We have developed a highly sensitive method for the analysis of deoxynucleotide monophosphates (dNMPs), which involves the use of liquid chromatography/mass spectrometry (LC/MS) and a new metal-free column. The new column solves the problem that the phosphate group in dNMPs