Skip to Content
Merck
All Photos(1)

Key Documents

COLO 320 DMF

93051118, human colon, Round & Refractile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human colon
growth mode:
Suspension
karyotype:
Not specified
morphology:
Round & Refractile
products:
Norepinephrine, epinephrine, serotonin, adrenocorticotropic, parathyroid hormones
receptors:
Not specified
relevant disease(s):
cancer
Pricing and availability is not currently available.

Product Name

COLO 320 DMF, 93051118

biological source

human colon

growth mode

Suspension

karyotype

Not specified

morphology

Round & Refractile

products

Norepinephrine, epinephrine, serotonin, adrenocorticotropic, parathyroid hormones

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colon carcinoma

Cell Line Description

Derived from a colon cancer biopsy of a 55 year old female.

Application

Tumourigenicity studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Will form clusters in suspension.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service