Recommended Products
Product Name
RENT tsS1.1, 92092515
biological source
rat endometrium
growth mode
Adherent
karyotype
Not specified
morphology
Cuboidal
products
Not specified
receptors
Glucocorticoid
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Rat Sprague-Dawley, primary endometrial, SV40 T antigen transformed
Cell Line Description
Derived from the endometrium tissue from the uterus of a 21-28 day-old Sprague-Dawley rat. The cells carry vimentin, CSF-1 (at 39°C), desmin, collagen type III and connexin 43 markers. The transforming agent is a ts mutant of SV 40 T-antigen (tsA58).
Culture Medium
DMEM + 20μg/l Gly-His-Lys (Sigma) + 2mM Glutamine + 80 IU human Insulin/l (Novo) + 10% Foetal Bovine Serum (FBS) (heat-inactivated).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 3x10,000 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Doubling time 48 hours. Cells show continuous growth at 34°C and growth arrest at 39°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service