Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Please contact Customer Service for Availability | 10 700,00 CZK |
About This Item
biological source
human coronary artery
packaging
pkg of 1,000,000 cells
morphology
(endothelial)
application(s)
cell analysis (DNA, RNA, and protein)
relevant disease(s)
hypertension; cardiovascular diseases
shipped in
dry ice
storage temp.
−20°C
General description
Application
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Other Notes
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description 1 million cells per pellet | description 500,000 proliferating cells | description 500,000 cryopreserved cells | description 500,000 proliferating cells |
| biological source human coronary artery | biological source human coronary artery | biological source human coronary artery | biological source human aorta (thoracic and abdominal) |
| morphology (endothelial) | morphology ( endothelial) | morphology ( endothelial) | morphology ( endothelial) |
| relevant disease(s) hypertension; cardiovascular diseases | relevant disease(s) - | relevant disease(s) - | relevant disease(s) hypertension; cardiovascular diseases |
| storage temp. −20°C | storage temp. room temp | storage temp. −196°C | storage temp. room temp |
| shipped in dry ice | shipped in ambient | shipped in dry ice | shipped in ambient |
| packaging pkg of 1,000,000 cells | packaging pkg of 500,000 cells | packaging pkg of 500,000 cells | packaging pkg of 500,000 cells |
Still not finding the right product?
Explore all of our products under Human Coronary Artery Endothelial Cells (HCAEC)
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
7783-20-2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

