Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Please contact Customer Service for Availability | CZK 4,990.00 |
About This Item
biological source
human foreskin (juvenile epidermis)
packaging
pkg of 1,000,000 cells
morphology
(epidermal)
application(s)
cell analysis (DNA, RNA, and protein)
shipped in
dry ice
storage temp.
−20°C
General description
Application
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Other Notes
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| morphology (epidermal) | morphology (epidermal) | morphology (epidermal) | morphology ( epidermal) |
| biological source human foreskin (juvenile epidermis) | biological source human epidermis (adult) | biological source human foreskin (juvenile epidermis) | biological source human foreskin (juvenile epidermis) |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in ambient |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. room temp |
| packaging pkg of 1,000,000 cells | packaging pkg of 1,000,000 cells | packaging pkg of 1,000,000 cells | packaging pkg of 500,000 cells |
| application(s) cell analysis (DNA, RNA, and protein) | application(s) cell analysis (DNA, RNA, and protein) | application(s) cell analysis (DNA, RNA, and protein) | application(s) - |
Still not finding the right product?
Explore all of our products under Normal Human Epidermal Keratinocytes (NHEK)
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
7783-20-2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

