Skip to Content
Merck

Skip To

C1312

Color Coders for Nalgene® cryogenic vials

yellow

Synonym(s):

vial cap inserts, vial identifier, Nalgene 5045-0002, colour coded insert, colour coded vial insert

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 ea
Please contact Customer Service for Availability
CZK 570.00

About This Item

NACRES:
NB.22
UNSPSC Code:
41103032
Manufacturer/tradename:
Nalgene 5045-0002
Material:
polystyrene

CZK 570.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

manufacturer/tradename

Nalgene 5045-0002

color

yellow

General description

Colored disks fit into the recessed tops of Nalgene cryogenic vials. The system allows color-coding of vials for quick visual identification. Cut out top facilitates handling of vials with forceps. Flat surface permits writing.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C0812C1062C0937
manufacturer/tradename

Nalgene 5045-0002

manufacturer/tradename

Nalgene 5045-0003

manufacturer/tradename

Nalgene 5045-0005

manufacturer/tradename

Nalgene 5045-0004

material

polystyrene

material

polystyrene

material

polystyrene

material

polystyrene

color

yellow

color

blue

color

red

color

green


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
C1312-100EA04061832614311