Skip to Content
Merck
All Photos(1)

Key Documents

C1119

Sigma-Aldrich

Caroverine hydrochloride

≥98% (HPLC), solid

Synonym(s):

1-[2-(Diethylamino)ethyl]-3-[(4-methoxyphenyl)methyl]-2(1H)-quinoxalinone monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28ClN3O2
CAS Number:
Molecular Weight:
401.93
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: >10 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

Cl.CCN(CC)CCN1C(=O)C(Cc2ccc(OC)cc2)=Nc3ccccc13

InChI

1S/C22H27N3O2.ClH/c1-4-24(5-2)14-15-25-21-9-7-6-8-19(21)23-20(22(25)26)16-17-10-12-18(27-3)13-11-17;/h6-13H,4-5,14-16H2,1-3H3;1H

InChI key

JRNWTJUIMRLKBV-UHFFFAOYSA-N

Biochem/physiol Actions

Caroverine hydrochloride is a nonselective NMDA and AMPA glutamate receptor antagonist.
Caroverine hydrochloride is a nonselective NMDA and AMPA glutamate receptor antagonist. Also, Caroverine hydrochloride is a class B calcium-channel-blocker, antiglutamatergic agent, and acts as an antioxidant when administered to lab animals.

Features and Benefits

This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Gedlicka et al.
American journal of otolaryngology, 30(3), 157-161 (2009-05-05)
This retrospective study was performed to evaluate the effectiveness of tympanotomy and sealing of the round window membrane after unilateral acute hearing loss. All patients presenting idiopathic sudden hearing loss, acoustic, or barotrauma were treated with prednisolone and caroverine. Thirty-six
S V Logvinov et al.
Morfologiia (Saint Petersburg, Russia), 133(1), 46-50 (2008-12-17)
The changes of interneuronal contacts in the internal reticular layer of albino rat retina were studied 7 and 30 days after the exposure to high intensity light (6000 Lux for 6 h). In osmicated preparations, the "light" type of synapse
Zhiqiang Chen et al.
Acta oto-laryngologica, 123(8), 905-909 (2003-11-11)
Intense sound stimulation may result in excessive glutamate release from the inner hair cells, resulting in binding to the postsynaptic glutamate receptors and leading to neuronal degeneration and functional impairment. In this study we investigated the therapeutic effect and time
Klaus Ehrenberger
The international tinnitus journal, 11(1), 34-37 (2006-01-20)
This prospective study, which conformed with good clinical practice (GCP-conform), tested the concept that the topical transtympanic administration of the quinoxaline derivative Caroverine promises a new approach to the treatment of tinnitus. The rationale for the study is the hypothesis
Dario Alpini et al.
The international tinnitus journal, 10(1), 91-93 (2004-09-24)
The difficulty of accurately localizing the source of subjective tinnitus is well-known. Anamnesis and traditional audiological tests can often suggest a source if its origin as peripheral or merely central (or both). Therefore, several authors, such as Risey, Denk, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service