Skip to Content
Merck

Skip To

C0295

Z-Pro-OSu

Synonym(s):

N-Cbz-L-proline N-hydroxysuccinimide ester, Z-L-proline hydroxysuccinimide ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C17H18N2O6
CAS Number:
Molecular Weight:
356.41
UNSPSC Code:
12352200
EC Number:
222-255-9
Beilstein/REAXYS Number:
500781
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

−20°C

SMILES string

O=C1CCC(=O)N1OC(=O)[C@@H]2CCCN2C(=O)OCc3ccccc3





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library